Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-tomoxetine hydrochloride | 82857-39-4 | MFCD11035876 | 99.8% | 291.82 g/mol | C17H22ClNO | 10 MG
SDP

Catalog No. 5000281882
Encompass_Preferred

(S)-Tomoxetine hydrochloride is the S-enantiomer of tomoxetine supplied as a white to off-white solid for research and analytical applications. It is an isomer of a norepinephrine reuptake inhibitor and is reported to inhibit norepinephrine reuptake with a Ki of 16.8 nM; not for human use.

  • High purity: 99.8%.
  • Appearance: white to off-white solid.
  • Reported activity: norepinephrine reuptake inhibition (Ki = 16.8 nM).
  • Solubility: DMSO 100 mg/mL (requires ultrasonic).
  • Recommended storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years).
  • Available pack sizes: 5 MG, 10 MG, larger sizes by quote.

Catalog No. 50-002-81882 Supplier Medchemexpress LLC Supplier No. HYW29742110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Tomoxetine hydrochloride is the S-enantiomer of tomoxetine supplied as a white to off-white solid for research and analytical applications. It is an isomer of a norepinephrine reuptake inhibitor and is reported to inhibit norepinephrine reuptake with a Ki of 16.8 nM; not for human use.

  • High purity: 99.8%.
  • Appearance: white to off-white solid.
  • Reported activity: norepinephrine reuptake inhibition (Ki = 16.8 nM).
  • Solubility: DMSO 100 mg/mL (requires ultrasonic).
  • Recommended storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years).
  • Available pack sizes: 5 MG, 10 MG, larger sizes by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.