Learn More
Medchemexpress LLC (S)-Tomoxetine hydrochloride | 82857-39-4 | MFCD11035876 | 99.8% | 291.82 g/mol | C17H22ClNO | 5 MG
SDP
Catalog No. 5000281864
Encompass_Preferred
(S)-Tomoxetine hydrochloride is the S-enantiomer of tomoxetine, a norepinephrine reuptake inhibitor used as a research reagent to study norepinephrine transport and related pharmacology. It is supplied as a high-purity hydrochloride salt in small, pre-weighed pack sizes for laboratory use; not for human or veterinary use.
  • High purity suitable for analytical and preclinical research.
  • Reported Ki of 16.8 nM for norepinephrine reuptake.
  • Molecular weight 291.82 g/mol; formula C17H22ClNO.
  • Provided with purity specification and batch traceability.
  • Available in small milligram pack sizes for assay work.
  • Intended for laboratory research use only.
Catalog No. 50-002-81864 Supplier Medchemexpress LLC Supplier No. HYW2974215MG
(S)-Tomoxetine hydrochloride is the S-enantiomer of tomoxetine, a norepinephrine reuptake inhibitor used as a research reagent to study norepinephrine transport and related pharmacology. It is supplied as a high-purity hydrochloride salt in small, pre-weighed pack sizes for laboratory use; not for human or veterinary use.
  • High purity suitable for analytical and preclinical research.
  • Reported Ki of 16.8 nM for norepinephrine reuptake.
  • Molecular weight 291.82 g/mol; formula C17H22ClNO.
  • Provided with purity specification and batch traceability.
  • Available in small milligram pack sizes for assay work.
  • Intended for laboratory research use only.