Learn More
Protifi LLC S-Trap mini kit (10x 100 – 300 ug)
SDP
Catalog No. NC1828287
Encompass
S-Trap sample processing is now available in a full kit format. The kit includes lysis, wash, and digestion buffers. The user must supply HPLC grade MeOH.
Catalog No. NC1828287 Supplier Protifi LLC Supplier No. K02MINI10
S-Trap sample processing is now available in a full kit format. The kit includes lysis, wash, and digestion buffers. The user must supply HPLC grade MeOH.