Learn More
Bon Opus Biosciences Recombinant S-trimer protein(Mammalian, C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences DRA4950UG
Encompass_Preferred
Recombinant 2019-nCoV S-trimer Protein is produced by our Mammalian expression system and the target gene encoding Cys15-Gln1208 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# QHD43416.1. Formulation: Supplied as a 0.2 μm filtered solution of PBS, PH 7.4. Bon Opus Cat# DRA49
Catalog No. 50-208-0540