Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

s-Trithiane 97%

Catalog No. AC140520250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC140520250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC140520250 Supplier Thermo Scientific Chemicals Supplier No. 140520250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 291-21-4 | C3H6S3 | 138.261 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291-21-4
Molecular Formula C3H6S3
Molecular Weight (g/mol) 138.261
MDL Number MFCD00006653
InChI Key LORRLQMLLQLPSJ-UHFFFAOYSA-N
Synonym s-trithiane, trithioformaldehyde, sym-trithian, thioform, trimethylene trisulfide, 1,3,5-trithiacyclohexane, trimethylentrisulfid, sym-trithiane, formaldehyde, thio-, trimer, thioform czech
PubChem CID 9264
ChEBI CHEBI:39196
IUPAC Name 1,3,5-trithiane
SMILES C1SCSCS1

Specifications

Assay Percent Range 97%
Quantity 25 g
Formula Weight 138.26
Chemical Name or Material s-Trithiane, 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.