Learn More
Medchemexpress LLC S-Trityl-L-cysteine | 2799-07-7 | 98.99% | 363.47 | 100 MG
SDP
Catalog No. 5000451871
Encompass_Preferred
S-Trityl-L-cysteine (NSC 83265) is a selective and allosteric kinesin Eg5 inhibitor, demonstrating an IC50 of 1 μM for the inhibition of basal ATPase activity and 140 nM for the microtubule-activated ATPase activity. This compound also exhibits antitumor activities.
  • Selective and allosteric kinesin Eg5 inhibitor.
  • Inhibits basal ATPase activity with an IC50 of 1 μM.
  • Inhibits microtubule-activated ATPase activity with an IC50 of 140 nM.
  • Possesses antitumor activities.
  • Binds to a unique pocket within the Eg5 motor domain.
  • Mediates cell apoptosis and cell cycle arrest in a dose-dependent manner (1-20 μM for 72 hours).
  • Triggers apoptosis and cell cycle arrest through the activation of mitogen-activated protein kinase and nuclear factor kB signaling pathways.
  • Induces mitotic arrest in HeLa cells (IC50 of 700 nM), leading to characteristic monoastral spindles.
Catalog No. 50-004-51871 Supplier Medchemexpress LLC Supplier No. HYW011102100MG
S-Trityl-L-cysteine (NSC 83265) is a selective and allosteric kinesin Eg5 inhibitor, demonstrating an IC50 of 1 μM for the inhibition of basal ATPase activity and 140 nM for the microtubule-activated ATPase activity. This compound also exhibits antitumor activities.
  • Selective and allosteric kinesin Eg5 inhibitor.
  • Inhibits basal ATPase activity with an IC50 of 1 μM.
  • Inhibits microtubule-activated ATPase activity with an IC50 of 140 nM.
  • Possesses antitumor activities.
  • Binds to a unique pocket within the Eg5 motor domain.
  • Mediates cell apoptosis and cell cycle arrest in a dose-dependent manner (1-20 μM for 72 hours).
  • Triggers apoptosis and cell cycle arrest through the activation of mitogen-activated protein kinase and nuclear factor kB signaling pathways.
  • Induces mitotic arrest in HeLa cells (IC50 of 700 nM), leading to characteristic monoastral spindles.