Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Trityl-L-cysteine 95.0+%, TCI America™
SDP

Catalog No. T25225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25225G Supplier TCI America Supplier No. T25225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A non-natural, sulfur-containing amino acid commonly used as a reagent in solution phase peptide synthesis.

Chemical Identifiers

CAS 2799-07-7
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
MDL Number MFCD00002611
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
Synonym s-trityl-l-cysteine, +-s-trityl-l-cysteine, h-cys trt-oh, s-tritylcysteine, tritylcysteine, 3-tritylthio-l-alanine, 2r-2-amino-3-triphenylmethyl sulfanyl propanoic acid, l-alanine, 3-tritylthio, stlc, s-triphenylmethyl-l-cysteine
PubChem CID 76044
IUPAC Name (2R)-2-amino-3-tritylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)N

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 5 g
Formula Weight 363.48
Percent Purity ≥95.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material S-Trityl-L-cysteine
RTECSNumber AY7710000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.