Learn More
Medchemexpress LLC HY-101445B 50mg Medchemexpress, (S)-Trolox CAS:53174-06-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101445B50MG
Encompass_Preferred
Medchemexpress, HY-101445B 50mg (S)-Trolox CAS:53174-06-4 (S)-Trolox is a analogue of vitamin E, in which the phytyl chain is replaced with a carboxyl group. (S)-Trolox is frequently used as a model compound for studies of structural features, as well as a standard for evaluation of antioxidant activity. (S)-Trolox has potent and specific neuroprotective and antioxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7850