Learn More
Creative Biomart Recombinant Salmonella Typhi HylE Antigen, His-tagged
Supplier:  Creative Biomart STH418
Encompass_Preferred
Recombinant S.Typhi HylE produced in E. coli is a non-glycosylated polypeptide chain having a molecular mass of 34 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-4796