Learn More
Seracare Life Sciences Inc S TYPHIMURIUM POS CONTROL
Catalog No. 5067420
Encompass_Preferred
Heat-killed Salmonella typhimurium cells. It consists of approximately 5 x 10^9 cells/mL (1 mL of 2% w/v wet packed cells) in dextran solution. It is intended for use as a positive control in immunoassays designed for Salmonella detection.
This item is not returnable. View return policy
Catalog No. 50-674-20 Supplier Seracare Life Sciences Inc Supplier No. 53700002
Heat-killed Salmonella typhimurium cells. It consists of approximately 5 x 10^9 cells/mL (1 mL of 2% w/v wet packed cells) in dextran solution. It is intended for use as a positive control in immunoassays designed for Salmonella detection.