Learn More
Medchemexpress LLC (S)-UFR2709 hydrochloride | 2934318-93-9 | 98.1% | 255.74 | 100 MG
SDP
Catalog No. 5000348527
Encompass_Preferred
(S)-UFR2709 hydrochloride is a competitive nAChR antagonist that displays higher affinity for α4β2 nAChRs than for α7 nAChRs. It decreases anxiety and reduces ethanol consumption and ethanol preference in alcohol-preferring rats. It acts as an anxiolytic agent and can be used for the study of nicotine addiction.
  • Competitive nAChR antagonist
  • Exhibits higher affinity for α4β2 nAChRs than for α7 nAChRs
  • Decreases anxiety
  • Reduces ethanol consumption and ethanol preference
  • Acts as an anxiolytic agent
  • Used for the study of nicotine addiction
  • Molecular target for the treatment of alcohol abuse and dependence
  • Does not affect the weight or locomotor activity of rats
Catalog No. 50-003-48527 Supplier Medchemexpress LLC Supplier No. HY137231B100MG
(S)-UFR2709 hydrochloride is a competitive nAChR antagonist that displays higher affinity for α4β2 nAChRs than for α7 nAChRs. It decreases anxiety and reduces ethanol consumption and ethanol preference in alcohol-preferring rats. It acts as an anxiolytic agent and can be used for the study of nicotine addiction.
  • Competitive nAChR antagonist
  • Exhibits higher affinity for α4β2 nAChRs than for α7 nAChRs
  • Decreases anxiety
  • Reduces ethanol consumption and ethanol preference
  • Acts as an anxiolytic agent
  • Used for the study of nicotine addiction
  • Molecular target for the treatment of alcohol abuse and dependence
  • Does not affect the weight or locomotor activity of rats