Learn More
Medchemexpress LLC (S)-UFR2709 hydrochloride | 2934318-93-9 | 98.1% | 255.74 | 50 MG
SDP
Catalog No. 5000351550
Encompass_Preferred
(S)-UFR2709 hydrochloride is a competitive nAChR antagonist that shows higher affinity for α4β2 nAChRs than for α7 nAChRs. It has been observed to decrease anxiety and reduce ethanol consumption and preference in alcohol-preferring rats. It acts as an anxiolytic agent and can be used for the study of nicotine addiction.
  • Competitive nAChR antagonist with higher affinity for α4β2 nAChRs than α7 nAChRs
  • Decreases anxiety
  • Reduces ethanol consumption and ethanol preference in alcohol-preferring rats
  • Acts as an anxiolytic agent
  • Can be used for the study of nicotine addiction
  • Inhibits nicotine reward
  • Decreases nicotine-evoked mRNA expression of α4 nACh receptor subunit in the brain of adult zebrafish
  • Does not affect the weight or locomotor activity of rats at effective doses for reducing alcohol consumption
Catalog No. 50-003-51550 Supplier Medchemexpress LLC Supplier No. HY137231B50MG
(S)-UFR2709 hydrochloride is a competitive nAChR antagonist that shows higher affinity for α4β2 nAChRs than for α7 nAChRs. It has been observed to decrease anxiety and reduce ethanol consumption and preference in alcohol-preferring rats. It acts as an anxiolytic agent and can be used for the study of nicotine addiction.
  • Competitive nAChR antagonist with higher affinity for α4β2 nAChRs than α7 nAChRs
  • Decreases anxiety
  • Reduces ethanol consumption and ethanol preference in alcohol-preferring rats
  • Acts as an anxiolytic agent
  • Can be used for the study of nicotine addiction
  • Inhibits nicotine reward
  • Decreases nicotine-evoked mRNA expression of α4 nACh receptor subunit in the brain of adult zebrafish
  • Does not affect the weight or locomotor activity of rats at effective doses for reducing alcohol consumption