Learn More
MSC Guardair Vac Generating Head, 1-1/2 in. Hose, 1 ea
Catalog No. 502867897
Encompass
The Guardair 10 gal. Vac Generating Head is designed for instant, high-performance vacuum capability when mounted on any Guardair 10 gal. canister. Powered by compressed air, it is dependable and trouble-free with no motors or moving parts, ideal for vacuuming chips, dust, debris, liquids, or sludges.
  • Vacuum generating head with exhaust silencer
  • Standard cartridge filter
  • Air shutoff valve with FNPT inlet and fittings
  • Produces exceptional vacuum lift and vacuum flow
  • Operates quietly and efficiently
  • Equipped with a medium vacuum lift (S venturi)
  • Ideal for wet/dry applications
Catalog No. 50-286-7897 Supplier MSC Supplier No. 39466578
The Guardair 10 gal. Vac Generating Head is designed for instant, high-performance vacuum capability when mounted on any Guardair 10 gal. canister. Powered by compressed air, it is dependable and trouble-free with no motors or moving parts, ideal for vacuuming chips, dust, debris, liquids, or sludges.
  • Vacuum generating head with exhaust silencer
  • Standard cartridge filter
  • Air shutoff valve with FNPT inlet and fittings
  • Produces exceptional vacuum lift and vacuum flow
  • Operates quietly and efficiently
  • Equipped with a medium vacuum lift (S venturi)
  • Ideal for wet/dry applications