Learn More
MSC Guardair Air Powered Wet/Dry Drum Vacuum Head, HEPA, 105 in Water Lift
Catalog No. 502838069
Encompass
This air-powered wet/dry drum vacuum head from Guardair is designed for industrial applications requiring efficient cleanup. It features HEPA filtration for effective particle capture and provides a strong 105 inches of water lift. Compatible with 1-1/2 inches hoses, it is ideal for use with 20-gallon drums for various wet and dry vacuuming tasks.
  • Air powered
  • Wet and dry drum vacuum head application
  • HEPA filtration
  • 105 in water lift
  • Compatible with 1-1/2 in hose
  • Compatible with 20 gal drum capacity
  • 95 dB sound level
  • Includes standard filter
Catalog No. 50-283-8069 Supplier MSC Supplier No. 39466560
This air-powered wet/dry drum vacuum head from Guardair is designed for industrial applications requiring efficient cleanup. It features HEPA filtration for effective particle capture and provides a strong 105 inches of water lift. Compatible with 1-1/2 inches hoses, it is ideal for use with 20-gallon drums for various wet and dry vacuuming tasks.
  • Air powered
  • Wet and dry drum vacuum head application
  • HEPA filtration
  • 105 in water lift
  • Compatible with 1-1/2 in hose
  • Compatible with 20 gal drum capacity
  • 95 dB sound level
  • Includes standard filter