Learn More
Medchemexpress LLC S-verapamil hydrochloride | 36622-28-3 | MFCD11045898 | 98.3% | 491.06 | C27H39ClN2O4 | 5 MG
SDP
Catalog No. 5000222561
Encompass_Preferred
(S)-Verapamil hydrochloride is the S-enantiomer of the calcium channel blocker verapamil, provided as a hydrochloride salt for research use. It functions as a P-glycoprotein and MRP1 inhibitor and is employed in in vitro pharmacology and drug transport studies, including investigations of multidrug resistance.
  • Single S-enantiomer for stereospecific pharmacology.
  • Hydrochloride salt form for improved solubility and handling.
  • Reported to inhibit P-gp and MRP1-mediated transport.
  • Suitable for in vitro assays and pharmacological studies.
  • Available as solid and DMSO solution formats.
Catalog No. 50-002-22561 Supplier Medchemexpress LLC Supplier No. HY135336A5MG
(S)-Verapamil hydrochloride is the S-enantiomer of the calcium channel blocker verapamil, provided as a hydrochloride salt for research use. It functions as a P-glycoprotein and MRP1 inhibitor and is employed in in vitro pharmacology and drug transport studies, including investigations of multidrug resistance.
  • Single S-enantiomer for stereospecific pharmacology.
  • Hydrochloride salt form for improved solubility and handling.
  • Reported to inhibit P-gp and MRP1-mediated transport.
  • Suitable for in vitro assays and pharmacological studies.
  • Available as solid and DMSO solution formats.