Learn More
Medchemexpress LLC S-verapamil hydrochloride | 36622-28-3 | MFCD11045898 | 98.3% | C27H39ClN2O4 | 10MG
SDP
Catalog No. 5000261745
Encompass_Preferred
(S)-Verapamil hydrochloride is the S-enantiomer of the calcium channel blocker verapamil, supplied as the hydrochloride salt for use in pharmacology and transporter research. It is reported to inhibit leukotriene C4 (LTC4) and calcein transport by the multidrug resistance protein MRP1 and is provided as a high-purity research reagent.
  • Enantiomer-specific activity useful for stereoselective studies.
  • Suitable for transporter and pharmacology assays.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Reported inhibition of LTC4 and MRP1-mediated transport.
  • High purity (98.3%) for reproducible experimental results.
Catalog No. 50-002-61745 Supplier Medchemexpress LLC Supplier No. HY135336A10MG
(S)-Verapamil hydrochloride is the S-enantiomer of the calcium channel blocker verapamil, supplied as the hydrochloride salt for use in pharmacology and transporter research. It is reported to inhibit leukotriene C4 (LTC4) and calcein transport by the multidrug resistance protein MRP1 and is provided as a high-purity research reagent.
  • Enantiomer-specific activity useful for stereoselective studies.
  • Suitable for transporter and pharmacology assays.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Reported inhibition of LTC4 and MRP1-mediated transport.
  • High purity (98.3%) for reproducible experimental results.