Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-viloxazine-d5 (hydrochloride) | 1246816-39-6 | 98.2% | 278.79 g·mol⁻1 | C13H15D5ClNO3 | 10MG
SDP

Supplier:  Medchemexpress LLC HY125784BS10MG

Encompass_Preferred

(S)-Viloxazine-d5 (hydrochloride) is the deuterium-labeled form of (S)-viloxazine hydrochloride supplied as the hydrochloride salt for use as a stable-isotope internal standard and research reagent. It is provided in small research quantities for analytical and assay applications.

  • Deuterium-labeled internal standard (d5) for analytical assays.
  • Hydrochloride salt form for improved handling.
  • High reported purity (98.19%).
  • Molecular weight approximately 278.79 g·mol⁻1.
  • Chemical formula includes five deuterium atoms (C13H15D5ClNO3).

Catalog No. 50-002-39085


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.