Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-viloxazine-d5 (hydrochloride) | 1246816-39-6 | 98.2% | 278.79 g/mol | C13H15D5ClNO3 | 1MG
SDP

Catalog No. 5000239086
Encompass_Preferred

(S)-Viloxazine-d5 (hydrochloride) is the deuterium-labeled hydrochloride salt of (S)-viloxazine, provided as an isotopically labeled reference standard for laboratory research and analytical quantification. It is intended for use as an internal standard in mass spectrometry and other analytical methods and is supplied as a solid hydrochloride salt for research use only.

  • Isotopically labeled internal standard for quantitative mass spectrometry.
  • Deuterated chemical formula C13H15D5ClNO3.
  • Molecular weight 278.79 g/mol.
  • Purity 98.2%.
  • Supplied as a hydrochloride salt in small milligram pack sizes.
  • Intended for research use only; not for human or therapeutic use.
  • Matches CAS number 1246816-39-6 for accurate substance identification.

Catalog No. 50-002-39086 Supplier Medchemexpress LLC Supplier No. HY125784BS1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Viloxazine-d5 (hydrochloride) is the deuterium-labeled hydrochloride salt of (S)-viloxazine, provided as an isotopically labeled reference standard for laboratory research and analytical quantification. It is intended for use as an internal standard in mass spectrometry and other analytical methods and is supplied as a solid hydrochloride salt for research use only.

  • Isotopically labeled internal standard for quantitative mass spectrometry.
  • Deuterated chemical formula C13H15D5ClNO3.
  • Molecular weight 278.79 g/mol.
  • Purity 98.2%.
  • Supplied as a hydrochloride salt in small milligram pack sizes.
  • Intended for research use only; not for human or therapeutic use.
  • Matches CAS number 1246816-39-6 for accurate substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.