Learn More
Medchemexpress LLC (S)-VQW-765 ((S)-AQW-051) | 98.0% | 294.39 | 5 MG
SDP
Catalog No. 5000401102
Encompass_Preferred
(S)-VQW-765 ((S)-AQW-051) is an orally active, selective, and effective α7 nicotinic ACh receptor (nAChR) partial agonist. It has potential applications in cognitive disorders related to neurological diseases, such as Alzheimer's disease or schizophrenia.
  • Orally active, selective, and effective α7 nAChR partial agonist
  • Potential applications in cognitive disorders
  • Suitable for neurological diseases like Alzheimer's or schizophrenia
Catalog No. 50-004-01102 Supplier Medchemexpress LLC Supplier No. HY19490A5MG
(S)-VQW-765 ((S)-AQW-051) is an orally active, selective, and effective α7 nicotinic ACh receptor (nAChR) partial agonist. It has potential applications in cognitive disorders related to neurological diseases, such as Alzheimer's disease or schizophrenia.
  • Orally active, selective, and effective α7 nAChR partial agonist
  • Potential applications in cognitive disorders
  • Suitable for neurological diseases like Alzheimer's or schizophrenia