Learn More
MSC S/W MAGNUM 3G GLASSES
Catalog No. 5000796273
Encompass
These safety glasses provide exceptional protection and comfort for various activities. They meet ANSI Z87.1+ standards and offer 99.9% UVA/UVB/UVC protection, making them suitable for demanding outdoor and industrial environments.
  • Frameless sunglasses design
  • Extendable temples with three different lengths for a customized fit
  • Self-adjusting nose pad with rubber fingers designed to prevent glasses from slipping
  • Hinge stops prevent temple tips from scratching lenses
  • Green mirror lenses provide glare reduction, ideal for outdoor use
  • Hard-coated polycarbonate lenses for scratch resistance and 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards for impact
Catalog No. 50-007-96273 Supplier MSC Supplier No. 00410464
These safety glasses provide exceptional protection and comfort for various activities. They meet ANSI Z87.1+ standards and offer 99.9% UVA/UVB/UVC protection, making them suitable for demanding outdoor and industrial environments.
  • Frameless sunglasses design
  • Extendable temples with three different lengths for a customized fit
  • Self-adjusting nose pad with rubber fingers designed to prevent glasses from slipping
  • Hinge stops prevent temple tips from scratching lenses
  • Green mirror lenses provide glare reduction, ideal for outdoor use
  • Hard-coated polycarbonate lenses for scratch resistance and 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards for impact