Learn More
Bellco Glass, Inc. The Bellco Mini Refrigerated Roll-In Incubator has 21 cubic feet of volume and is designed for use with Bellco's Cell Production Roller Apparatus (CPRA) up to 2 decks high. The flush-to-floor design allows easy movement of the CPRA in and out of the cham
Supplier: Bellco Glass, Inc. 772804115
The Bellco Mini Refrigerated Roll-In Incubator has 21 cubic feet of volume and is designed for use with Bellco's Cell Production Roller Apparatus (CPRA) up to 2 decks high. The flush-to-floor design allows easy movement of the CPRA in and out of the cham