Learn More
Bellco Glass, Inc. The Bellco Mini Refrigerated Roll-In Incubator has 21 cubic feet of volume and is designed for use with Bellco's Cell Production Roller Apparatus (CPRA) up to 2 decks high. The flush-to-floor design allows easy movement of the CPRA in and out of the cham
SDP
Catalog No. 501946039
Encompass_Preferred
The Bellco Mini Refrigerated Roll-In Incubator has 21 cubic feet of volume and is designed for use with Bellco's Cell Production Roller Apparatus (CPRA) up to 2 decks high. The flush-to-floor design allows easy movement of the CPRA in and out of the cham
Catalog No. 50-194-6039 Supplier Bellco Glass, Inc. Supplier No. 772804230
Add to Cart
Add to Cart
Request a Quote
The Bellco Mini Refrigerated Roll-In Incubator has 21 cubic feet of volume and is designed for use with Bellco's Cell Production Roller Apparatus (CPRA) up to 2 decks high. The flush-to-floor design allows easy movement of the CPRA in and out of the cham