Learn More
Bellco Glass, Inc. Single Window Refridgerated Incubator, 115V
SDP
Catalog No. 501946033
Encompass_Preferred
The Bellco Refrigerated Roll-In Incubators have 40 cubic feet of volume. Refrigerated roll-in incubator ideal for insect cell incubations performed at 27 degrees c. Microprocessor PID temperature controller accommodates up to 9-deck fixed frame or base +7 modular Bellco cell production roller apparatus. Accessory power outlets (4) in chamber provided for supplemental equipment. Forced air circulation facilitates even temperature distribution. Control panel w/power switch, convenience outlet, safety thermostat refrigeration switch, temperature controller. "flush to floor' design allows easy loading of Bellco roller apparatus. Btu/hr: 5,120 max. MADE IN THE USA
Catalog No. 50-194-6033 Supplier Bellco Glass, Inc. Supplier No. 772801005
Add to Cart
Add to Cart
Request a Quote
The Bellco Refrigerated Roll-In Incubators have 40 cubic feet of volume. Refrigerated roll-in incubator ideal for insect cell incubations performed at 27 degrees c. Microprocessor PID temperature controller accommodates up to 9-deck fixed frame or base +7 modular Bellco cell production roller apparatus. Accessory power outlets (4) in chamber provided for supplemental equipment. Forced air circulation facilitates even temperature distribution. Control panel w/power switch, convenience outlet, safety thermostat refrigeration switch, temperature controller. "flush to floor' design allows easy loading of Bellco roller apparatus. Btu/hr: 5,120 max. MADE IN THE USA