Learn More
Abcam (S)-WAY 100135, 5-HT1A receptor antagonist, 50MG
Catalog No. 502704484
Encompass_Preferred
MW 468.5 Da, Purity >99%. Potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4484 Supplier Abcam Supplier No. AB14682150MG
MW 468.5 Da, Purity >99%. Potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement