Learn More
Medchemexpress LLC (S)-WSD0628 | 2765449-10-1 | 99.8% | 439.46 | 25 MG
SDP
Catalog No. 5000394304
Encompass_Preferred
(S)-WSD0628 (Compound 28) is the S-isomer of WSD0628. It is an inhibitor for ATM that inhibits the phosphorylation of ATM in MCF-7 cells with an IC50 <100 nM. (S)-WSD0628 exhibits radiosensitizing activity and can cross the blood-brain barrier.
  • ATM inhibitor
  • Inhibits phosphorylation of ATM in MCF-7 cells with IC50 <100 nM
  • Exhibits radiosensitizing activity
  • Can cross the blood-brain barrier
Catalog No. 50-003-94304 Supplier Medchemexpress LLC Supplier No. 25MGHY162002A
(S)-WSD0628 (Compound 28) is the S-isomer of WSD0628. It is an inhibitor for ATM that inhibits the phosphorylation of ATM in MCF-7 cells with an IC50 <100 nM. (S)-WSD0628 exhibits radiosensitizing activity and can cross the blood-brain barrier.
  • ATM inhibitor
  • Inhibits phosphorylation of ATM in MCF-7 cells with IC50 <100 nM
  • Exhibits radiosensitizing activity
  • Can cross the blood-brain barrier