Learn More
Medchemexpress LLC (S)-XY-05 | 3024030-10-9 | 99.8% | 447.41 g·mol⁻¹ | C20H20F3N7O2 | 10 MG
SDP
Catalog No. 5000288524
Encompass_Preferred
PARP7-IN-17 is a potent small-molecule inhibitor of PARP7 with an IC50 of 4.5 nM. It demonstrates oral bioavailability and reported antitumor activity. Supplied as a white to off-white solid for research use, it is commonly used in cellular and in vivo studies of PARP7 biology and anticancer mechanisms.
  • Potent PARP7 inhibition (IC50 4.5 nM).
  • Reported oral bioavailability and antitumor activity.
  • High purity (≈99.8%) suitable for research applications.
  • Solid, white to off-white appearance for ease of handling.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small milligram pack sizes for preclinical studies.
Catalog No. 50-002-88524 Supplier Medchemexpress LLC Supplier No. HY16308110MG
PARP7-IN-17 is a potent small-molecule inhibitor of PARP7 with an IC50 of 4.5 nM. It demonstrates oral bioavailability and reported antitumor activity. Supplied as a white to off-white solid for research use, it is commonly used in cellular and in vivo studies of PARP7 biology and anticancer mechanisms.
  • Potent PARP7 inhibition (IC50 4.5 nM).
  • Reported oral bioavailability and antitumor activity.
  • High purity (≈99.8%) suitable for research applications.
  • Solid, white to off-white appearance for ease of handling.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small milligram pack sizes for preclinical studies.