Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-XylBINAP 97.0+%, TCI America™
SDP

Catalog No. X00711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00711G 1 g
X0071200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00711G Supplier TCI America Supplier No. X00711G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135139-00-3
Molecular Formula C52H48P2
Molecular Weight (g/mol) 734.904
MDL Number MFCD01630821
InChI Key MXGXXBYVDMVJAO-UHFFFAOYSA-N
Synonym s-dm-binap, dm-binap, r-dm-binap, s---2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, s-3,5-xylyl-binap, r-+-2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, 3,5-xylyl-binap, s-xylyl-binap, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-1,1'-binaphthalene, r-+-xylbinap
PubChem CID 4189905
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C

Specifications

Melting Point 255°C
Color White-Yellow
Quantity 1 g
Formula Weight 734.90
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-XylBINAP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.