Learn More
Medchemexpress LLC S-ZINC-3573 10mM 1mL | 2095596-11-3 | 307.39 g·mol⁻¹ | C18H21N5 | 1 ML
SDP
Catalog No. 5000226286
Encompass_Preferred
(S)-ZINC-3573 is the S-enantiomer of ZINC-3573 used as an inactive control in studies of Mas-related G-protein-coupled receptor X2 (MRGPRX2) pharmacology. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solid quantities for in vitro research. Purity is reported at 99.89% and analytical files are provided for quality and safety.
Catalog No. 50-002-26286 Supplier Medchemexpress LLC Supplier No. HY11568210MM 1ML
(S)-ZINC-3573 is the S-enantiomer of ZINC-3573 used as an inactive control in studies of Mas-related G-protein-coupled receptor X2 (MRGPRX2) pharmacology. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solid quantities for in vitro research. Purity is reported at 99.89% and analytical files are provided for quality and safety.