Learn More
Bon Opus Biosciences Recombinant S1 Protein(Mammalian, C-10His) -50ug
SDP
Supplier:  Bon Opus Biosciences DRA5650UG
Encompass_Preferred
Recombinant 2019-nCoV S1 Protein is produced by our Mammalian expression system and the target gene encoding Gln14-Arg685 is expressed with a 10His tag at the C-terminus. Expression system: Human Cells. Accession# QHD43416.1. Formulation: Supplied as a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# DRA56
Catalog No. 50-208-0548