Learn More
Perkin Elmer US LLC S10 Autosampler Dust Cover
Catalog No. NC1809527
Encompass_Preferred
Plexiglass cover for reduced contamination of samples from ambient particulates. Measurements: 48cm (18.89) wide 61cm (24.01) high 39cm (15.35) deep.
Catalog No. NC1809527 Supplier Perkin Elmer US LLC Supplier No. N2020045
Plexiglass cover for reduced contamination of samples from ambient particulates. Measurements: 48cm (18.89) wide 61cm (24.01) high 39cm (15.35) deep.