Learn More
Sigma Aldrich Fine Chemicals Biosciences LpDNPH S10 Cartridge bed wt. 350 mg, volume 3 mL, pkg of 50 ea |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 21014
Encompass
LpDNPH S10 Cartridge bed wt. 350 mg, volume 3 mL, pkg of 50 ea |
Catalog No. NC0619236