Learn More
Genscript Corporation S100 2BD1 mAb Mouse
Catalog No. 5000057340
Encompass
S100 protein family contains variety of members such as homodimer and heterodimer It is a useful marker for monitoring brain damage and detecting secondary neurological injuries The concentration of S100 A8/A9 hetero complex increases in blood when special diseases take places such as rheumatoid arthritis cystic fibrosis multiple scerlosis and HIV infections S100A8/A9 protein is a potential serum marker for diagnosis of pancreatic ductal adenocarcinoma (PDAC) intraductal papillary mucinous neoplasm (IPMN) or chronic pancreatitis (CP)
Catalog No. 50-000-57340 Supplier Genscript Corporation Supplier No. V01002100
Add to Cart
Add to Cart
Request a Quote
S100 protein family contains variety of members such as homodimer and heterodimer It is a useful marker for monitoring brain damage and detecting secondary neurological injuries The concentration of S100 A8/A9 hetero complex increases in blood when special diseases take places such as rheumatoid arthritis cystic fibrosis multiple scerlosis and HIV infections S100A8/A9 protein is a potential serum marker for diagnosis of pancreatic ductal adenocarcinoma (PDAC) intraductal papillary mucinous neoplasm (IPMN) or chronic pancreatitis (CP)