Learn More
Alpco Ltd S100 Beta ELISA, 96 Wells, 20 L
SDP
Supplier:  Alpco Ltd 48CBPHUE01
Encompass_Preferred
S100 Beta ELISA is for the quantitative determination of S-100? in mouse, rat, and human plasma samples as well as rat and human cerebrospinal fluid (CSF) samples. For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-305-053