Learn More
Pyramex™ S1000 Medical Shield
Catalog No. 19817378 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
Medical Shield
Replacement Shield
Includes:
20 Shields
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-817-378 Medical Shield -
19-817-380 Replacement Shield 20 Shields
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-817-378 Supplier Pyramex™ Supplier No. S1000
  • $6.49
    $1.00 / Each of 1
    Save $5.49
    85% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
S1000 Medical Shield offers over the top face protection with polycarbonate face shield, foam headband, and adjustable strap.
  • Comfortable and functional over the top face protection
  • Adjustable head strap holds face shield in place
  • Polycarbonate face shield protects against splashes
  • Foam headband provides comfort
  • Can be worn over eyewear and face masks
  • Replacement faceshields are also available
Specifications
Dimensions (L x W) 302 x 215 mm
Lens Coating Scratch-resistant
Color Clear
Lens Tint Clear
Product Type Medical Shield
Headband Material Polypropylene
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Autoclavable Not Autoclavable