Learn More
Pyramex™ S1000 Medical Shield
Catalog No. 19817380 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
Medical Shield
Replacement Shield
Includes:
20 Shields
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-817-380 Replacement Shield 20 Shields
19-817-378 Medical Shield -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-817-380 Supplier Pyramex™ Supplier No. S1000R
Add to Cart
Add to Cart
This item is not returnable. View return policy
S1000 Medical Shield offers over the top face protection with polycarbonate face shield, foam headband, and adjustable strap.
  • Comfortable and functional over the top face protection
  • Adjustable head strap holds face shield in place
  • Polycarbonate face shield protects against splashes
  • Foam headband provides comfort
  • Can be worn over eyewear and face masks
  • Replacement faceshields are also available
Specifications
Dimensions (L x W) 302 x 215 mm
Lens Coating Scratch-resistant
Color Clear
Lens Tint Clear
Product Type Replacement Shield
Includes 20 Shields
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Autoclavable Not Autoclavable