Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6271MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 4C4.9. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a, kappa. Positive Control: Brain, Melanoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified bovine brain S100 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 10-12kDa.Chromosome: 1q21.3. Gene Symbol: S100A1. Gene ID: 6271.
Catalog No. 50-188-5149