Learn More
ABclonal Technology S100A12/CGRP Rabbit mAb
SDP
Catalog No. 5000005013
Encompass_Preferred
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs S100 proteins are localized in the cytoplasm and or nucleus of a wide range of cells and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation S100 genes include at least 13 members which are located as a cluster on chromosome 1q21 This protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities The protein includes an antimicrobial peptide which has antibacterial activity
Catalog No. 50-000-05013 Supplier ABclonal Technology Supplier No. A2391220UL
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs S100 proteins are localized in the cytoplasm and or nucleus of a wide range of cells and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation S100 genes include at least 13 members which are located as a cluster on chromosome 1q21 This protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities The protein includes an antimicrobial peptide which has antibacterial activity