Learn More
ACROBiosystems Recombinant Protein;Human S100A14 Protein;E.coli;100UG;S14,S100A14,S114,S100A15,S100-A14,S100-A15
Supplier: ACROBiosystems S14H5121100UG
Recombinant Protein;Human S100A14 Protein;100UG;Human S100A14, His Tag (S14-H5121) is expressed from E.coli cells. It contains AA Met 1 - His 104 (Accession # AAH05019).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.5 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).