Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human S100A14 Protein, His Tag, 1 mg

Catalog No. 501658799
Encompass_Preferred

Human S100A14 protein, containing a C-terminal polyhistidine tag, is expressed in E. coli cells. S100A14 is an S-100 family calcium-binding protein that acts as a homodimer and interacts with the receptor for advanced glycation end products (RAGE). It modulates P53/TP53 protein levels to regulate cell survival, apoptosis, and migration through MMP2 modulation.

  • Purity: >95% as determined by SDS-PAGE
  • Calculated MW: 12.5 kDa; migrates as 13 kDa under reducing conditions
  • Formulation: Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 8.0 with trehalose
  • Contains amino acids Met 1 to His 104 (Accession # Q9HCY8-1)
  • Storage: Stable for 12 months at -20°C to -70°C in lyophilized state; 3 months at -70°C after reconstitution
  • Avoid repeated freeze-thaw cycles

Catalog No. 50-165-8799 Supplier ACROBiosystems Supplier No. S14H51211MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human S100A14 protein, containing a C-terminal polyhistidine tag, is expressed in E. coli cells. S100A14 is an S-100 family calcium-binding protein that acts as a homodimer and interacts with the receptor for advanced glycation end products (RAGE). It modulates P53/TP53 protein levels to regulate cell survival, apoptosis, and migration through MMP2 modulation.

  • Purity: >95% as determined by SDS-PAGE
  • Calculated MW: 12.5 kDa; migrates as 13 kDa under reducing conditions
  • Formulation: Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 8.0 with trehalose
  • Contains amino acids Met 1 to His 104 (Accession # Q9HCY8-1)
  • Storage: Stable for 12 months at -20°C to -70°C in lyophilized state; 3 months at -70°C after reconstitution
  • Avoid repeated freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.