Learn More
Sino Biological S100A15 / S100A7A Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 50226RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse S100A15 / S100A7A (Catalog#50226-M10E; Q6S5I3; Met1-Tyr108). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7992