Learn More
Sino Biological S100A16 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11137RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A16 (rh S100A16; Catalog#11137-HNAE; Q96FQ6; Met 1-Ser 103). S100A16 specific IgG was purified by Human S100A16 affinity chromatography.
Catalog No. 50-224-8079