Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S100A3 protein, human (His-MBP) | 96.1% | 50 UG
SDP

Catalog No. 5000290418
Encompass_Preferred

Recombinant human S100A3 protein with an N-terminal His-MBP fusion tag, expressed in Escherichia coli and supplied as a 50 μg aliquot. The protein is formulated in PBS with 20% glycerol, 0.22 μm filtered, and has an endotoxin level <1 EU/μg. Intended for research use in biochemical and biophysical studies of S100 family proteins.

  • Recombinant human S100A3 with N-terminal His-MBP fusion tag.
  • Expressed in Escherichia coli for research-grade production.
  • Supplied in PBS, 20% glycerol, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg, suitable for most in vitro assays.
  • Approximate molecular weight ~50 kDa including fusion tag.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-90418 Supplier Medchemexpress LLC Supplier No. HYP7658550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human S100A3 protein with an N-terminal His-MBP fusion tag, expressed in Escherichia coli and supplied as a 50 μg aliquot. The protein is formulated in PBS with 20% glycerol, 0.22 μm filtered, and has an endotoxin level <1 EU/μg. Intended for research use in biochemical and biophysical studies of S100 family proteins.

  • Recombinant human S100A3 with N-terminal His-MBP fusion tag.
  • Expressed in Escherichia coli for research-grade production.
  • Supplied in PBS, 20% glycerol, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg, suitable for most in vitro assays.
  • Approximate molecular weight ~50 kDa including fusion tag.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.