Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6275MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: S100A4/1482. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa, A549 or A375 Cells. Placenta. Cellular Staining: Cytoplasmic and Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human S100A4 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 10-12kDa.Chromosome: 1q21. Gene Symbol: S100A4. Gene ID: 6275. Ensemble: ENSG00000196154. Uniprot: NP_062427.
Catalog No. 50-188-5151