Learn More
Aviva Systems Biology Corp S100A8 Antibody - middle region (ARP61367_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61367_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against S100A8. It was validated on Western Blot. . Alternate names: 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-287-355