Learn More
Medchemexpress LLC S100A8 Human N-His 500ug
SDP
Catalog No. 5000184157
Encompass_Preferred
Recombinant human S100A8 protein with an N-terminal His purification tag, provided as a 500 μg pack for biochemical and cell-based research. S100A8 is a calcium-binding protein involved in inflammatory and immune responses; the recombinant preparation is intended for studies of protein interactions, signaling pathways, and assay development. Verify purity and storage instructions before experimental use.
  • Recombinant human S100A8 with N-terminal His tag.
  • Supplied as 500 μg for flexible experimental use.
  • Suitable for biochemical assays, binding studies, and signaling research.
  • Produced for in vitro applications; validate performance in your assay.
  • Store according to supplier recommendations to maintain activity.
Catalog No. 50-001-84157 Supplier Medchemexpress LLC Supplier No. HYP70531A500UG
Recombinant human S100A8 protein with an N-terminal His purification tag, provided as a 500 μg pack for biochemical and cell-based research. S100A8 is a calcium-binding protein involved in inflammatory and immune responses; the recombinant preparation is intended for studies of protein interactions, signaling pathways, and assay development. Verify purity and storage instructions before experimental use.
  • Recombinant human S100A8 with N-terminal His tag.
  • Supplied as 500 μg for flexible experimental use.
  • Suitable for biochemical assays, binding studies, and signaling research.
  • Produced for in vitro applications; validate performance in your assay.
  • Store according to supplier recommendations to maintain activity.