Learn More
Medchemexpress LLC S100A8-S100A9 Hetero 20ug
SDP
Catalog No. 5000185533
Encompass_Preferred
Recombinant mouse S100A8-S100A9 heterodimer protein (His-tagged), expressed in E. coli and purified for use in biochemical and immunological studies of inflammation and innate immunity. The protein is characterized by defined molecular weight bands on SDS-PAGE and supplied in a small-format vial suitable for low-volume assays.
  • High purity suitable for biochemical and immunological assays.
  • N-terminal 6xHis tag for purification and detection.
  • Derived from mouse sequences for species-relevant studies.
  • Expressed in E. coli for reproducible recombinant production.
  • Provided in a 20 μg size for low-volume experiments.
Catalog No. 50-001-85533 Supplier Medchemexpress LLC Supplier No. HYP70543820UG
Recombinant mouse S100A8-S100A9 heterodimer protein (His-tagged), expressed in E. coli and purified for use in biochemical and immunological studies of inflammation and innate immunity. The protein is characterized by defined molecular weight bands on SDS-PAGE and supplied in a small-format vial suitable for low-volume assays.
  • High purity suitable for biochemical and immunological assays.
  • N-terminal 6xHis tag for purification and detection.
  • Derived from mouse sequences for species-relevant studies.
  • Expressed in E. coli for reproducible recombinant production.
  • Provided in a 20 μg size for low-volume experiments.