Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S100A8-S100A9 Hetero 500ug
SDP

Catalog No. 5000196562
Encompass_Preferred

Recombinant human S100A8-S100A9 heterodimer supplied as a His-tagged solution and expressed in E. coli. The calcium- and zinc-bound complex is involved in inflammatory and immune responses, contributes to leukocyte function, regulates the cytoskeleton, and can activate intracellular NADPH oxidase. Provided in a research-grade formulation for biochemical and cell-based assays.

  • Recombinant human heterodimer expressed in E. coli.
  • Includes His tag for detection and purification.
  • Contains calcium- and zinc-bound S100A8 subunit.
  • Suitable for biochemical and cell-based inflammation studies.
  • Supplied as a stabilized solution for easy handling.

Catalog No. 50-001-96562 Supplier Medchemexpress LLC Supplier No. HYP71076500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human S100A8-S100A9 heterodimer supplied as a His-tagged solution and expressed in E. coli. The calcium- and zinc-bound complex is involved in inflammatory and immune responses, contributes to leukocyte function, regulates the cytoskeleton, and can activate intracellular NADPH oxidase. Provided in a research-grade formulation for biochemical and cell-based assays.

  • Recombinant human heterodimer expressed in E. coli.
  • Includes His tag for detection and purification.
  • Contains calcium- and zinc-bound S100A8 subunit.
  • Suitable for biochemical and cell-based inflammation studies.
  • Supplied as a stabilized solution for easy handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.