Learn More
Medchemexpress LLC S100A8-S100A9 Hetero 50ug
SDP
Catalog No. 5000184329
Encompass_Preferred
Recombinant human S100A8-S100A9 heterodimer protein supplied as a purified solution with a C-terminal His tag. Produced by coexpression in E. coli and purified to >95% purity, the heterodimer (calprotectin) is suitable for biochemical and cell-based assays investigating inflammation, immune signaling, and metal-binding activity.
  • High purity (>95%) suitable for analytical and functional studies.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli for scalable production.
  • Available in multiple sizes including 50 μg for small-scale assays.
  • Contains human S100A8 and S100A9 sequences (UniProt P05109, P06702).
Catalog No. 50-001-84329 Supplier Medchemexpress LLC Supplier No. HYP70543850UG
Recombinant human S100A8-S100A9 heterodimer protein supplied as a purified solution with a C-terminal His tag. Produced by coexpression in E. coli and purified to >95% purity, the heterodimer (calprotectin) is suitable for biochemical and cell-based assays investigating inflammation, immune signaling, and metal-binding activity.
  • High purity (>95%) suitable for analytical and functional studies.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli for scalable production.
  • Available in multiple sizes including 50 μg for small-scale assays.
  • Contains human S100A8 and S100A9 sequences (UniProt P05109, P06702).