Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S100A8-S100A9 Hetero 5ug
SDP

Catalog No. 5000196477
Encompass_Preferred

Recombinant human S100A8-S100A9 heterodimer (His) supplied as a filtered protein solution for research use. Produced in E. coli and provided with a C-terminal 6xHis tag, the heterodimer (calprotectin) is intended for in vitro studies of inflammation and immune signaling; consult the lot-specific COA for exact formulation and storage conditions.

  • Recombinant human heterodimer expressed in E. coli.
  • C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in multiple pack sizes, including a 5 μg option.
  • Approximate molecular weights: ~13 kDa and 15-17 kDa by SDS-PAGE.
  • Supplied in 0.22 μm filtered buffered formulations; check COA for exact buffer.

Catalog No. 50-001-96477 Supplier Medchemexpress LLC Supplier No. HYP710765UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human S100A8-S100A9 heterodimer (His) supplied as a filtered protein solution for research use. Produced in E. coli and provided with a C-terminal 6xHis tag, the heterodimer (calprotectin) is intended for in vitro studies of inflammation and immune signaling; consult the lot-specific COA for exact formulation and storage conditions.

  • Recombinant human heterodimer expressed in E. coli.
  • C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in multiple pack sizes, including a 5 μg option.
  • Approximate molecular weights: ~13 kDa and 15-17 kDa by SDS-PAGE.
  • Supplied in 0.22 μm filtered buffered formulations; check COA for exact buffer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.