Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6280MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: S100A9/1011. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Liver or histiocytoma. Cellular Staining: Cytoplasmic, Extracellular (secreted), Cell Surface. Buffer: 10mM PBS with 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human S100A9 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 14kDa.Chromosome: 1q21. Gene Symbol: S100A9. Gene ID: 6280. Ensemble: ENSG00000163220. Uniprot: NP_002956.
Catalog No. 50-188-5152